Learn More
Thermo Scientific™ National Preassembled Target DP™ Kits
Product Code. p-4526330
Click to view available options
Closure Color:
Blue
Red
Septum:
PTFE/Rubber
PTFE/Silicone
PTFE/Silicone/PTFE
Color:
Amber
Clear
Unit Size:
Pack of 100
Product Code. 11901593
Brand: Thermo Scientific™ C4000180W
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Thermo Scientific™ National Preassembled Target DP Kits include 100 vials with pre-attached caps and septa.
Thermo Scientific™ National Preassembled Target DP Kits include 100 vials with pre-attached caps and septa.
  • Packaged in convenient vial trays with clear covers or in polybags
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Blue
Color (Seal) Red
Description PTFE/red rubber septa; Clear glass I-D
Color Clear
Material Glass
Volume (Metric) 2 mL
Septum PTFE/Rubber
Closure Size 9-425
Marking Spot Yes