Learn More
Thermo Scientific™ National Unassembled Certified™ Vial Kits
Thermo Scientific™ National Unassembled Certified Vial Kits provide extra assurance that time after time, the product upholds rigorous manufacturing standards.
Brand: Thermo Scientific™ CERT4000-71LV
Additional Details : Weight : 0.80000kg
Description
Thermo Scientific™ National Unassembled Certified Vial Kits provide extra assurance that time after time, the product upholds rigorous manufacturing standards.
- Fully lot tested for HPLC and GC analysis
- Certificate of Conformance in every pack
- Bonded caps to prevent septa push-through
- Clear Type 1, 33 expansion borosilicate glass or amber Type 2, 51 expansion borosilicate glass
- Available with or without ID patch
- Fixed micro vials or inserts for limited-volume sampling
Specifications
Certified Target DP 350µL Clear Glass I-D Fused Insert Vial, 9mm Screw Thread | |
PTFE, Silica Glass | |
350 μL | |
Bonded PTFE, Silicone | |
9-425 |
Clear | |
Yes | |
PTFE/Silica | |
Screw Cap | |
Yes |