missing translation for 'onlineSavingsMsg'
Learn More

Ark Plas Products™ Natural Polypropylene Commercial Grade Threaded Straight Connectors

Use these fittings for a wide range of applications. Ark Plas Products™ Natural Polypropylene Commercial Grade Threaded Straight Connectors are designed as a low cost alternative with high end performance.

Brand:  Ark Plas Products™ CGA23-PR0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 12673780

  • 51.86€ / Pack of 100


Estimated Shipment
14-05-2021
Add to basket

Description

Description

  • Economic and reliable
  • Up to 125psi without clamp
  • Contain recycled runner material
  • Available in Natural Nylon or Natural Polypropylene
Specifications

Specifications

Natural
6.4 mm
Threaded Straight Connector
1/4 in. Barb
Straight
Autoclavable
Polypropylene
25 Pack
1/4-18 UNF Male Thread
Videos
SDS
Documents