Learn More
Saint Gobain Life Sciences Natural Rubber Folding Skirt Stoppers
Use these stoppers for extra protection of tube ends during transport, handling and other operations during which tubes require both internal and end protection. Saint Gobain Life Sciences™ Natural Rubber Folding Skirt Stoppers provide an excellent seal.
Brand: Saint Gobain Life Sciences 407030-05
Additional Details : Weight : 0.10800kg
Description
- Stoppers are flexible and provide a complete seal and good adhesion
- Grey-blue grade: made from natural rubber
- Temperature resistance from -35 to +70°C (120°C in moist heat)
Specifications
Folding Skirt Stopper | |
22mmH/27.5mmH | |
1/Ea. |
30.7D | |
30.7mm | |
Natural Rubber |