Learn More
Saint Gobain Life Sciences Natural Rubber Stoppers
Click to view available options
Diameter (Metric) Bottom:
4 mm
5 mm
8 mm
10.5 mm
15 mm
17 mm
18 mm
21 mm
29 mm
34 mm
36 mm
41 mm
47 mm
Quantity:
10/Pk.
100/ck.
20/Pk.
5/Pk.
50/Pk.
Unit Size:
Pack of 10
Pack of 100
Pack of 20
Pack of 5
Pack of 50
Description
- Temperature resistance from -35 to +70°C (120°C in humid conditions)
- DIN Standard 18271
- Gray-blue color
Specifications
Specifications
| Height (Metric) | 40mm |
| Quantity | 10/Pk. |
| Color | Blue |
| Diameter (Metric) Bottom | 47 mm |
| Diameter (Metric) Top | 55mm |
| Material | Natural Rubber |
| Product Type | Stopper |
| Stopper Number | 54D |