Learn More
Deutsch Neumann™ Vacuum Quality Laboratory Tubing
Click to view available options
Diameter (Metric) Inner:
7 mm
Unit Size:
1m
Description
- Durometer 50 Shore A
- Good elastic memory
- Excellent choice for vacuum applications
Specifications
Specifications
| Diameter (Metric) Inner | 7 mm |
| Material | Natural Rubber |
| Diameter (Metric) Outer | 17 mm |
| Overall Length (Metric) | 25 m |
| Thickness (Metric) | 5 mm |
| Color | Red |