Learn More
Abeba™ Nature 4045 Shoes
Product Code. 15681328
Change view
Click to view available options
Size:
34
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15681328 34 1 pair
15691328 35 1 pair
15601338 36 1 pair
15611338 37 1 pair
15621338 38 1 pair
15631338 39 1 pair
15641338 40 1 pair
15651338 41 1 pair
15661338 42 1 pair
15671338 43 1 pair
15681338 44 1 pair
15691338 45 1 pair
15601348 46 1 pair
15611348 47 1 pair
15621348 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 15681328 Supplier Abeba™ Supplier No. 4045/34
Only null left
Add to basket
Add to basket
Leather covered cork footbed is ESD dissipative.
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather, perforated
  • Night blue
  • Slip resistance special rubber outsole sole
  • Leather coated cork footbed
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • Adjustable instep area
  • Buckle fastening
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, FO SRC
Color Night Blue
Footbed Material Cork
Material Leather
Size 34
Sole Features Slip-resistant
Sole Material Rubber
Style 4045