Learn More
Abeba™ Nature 4090 Shoes
Leather covered cork footbed is ESD dissipative.
81.95€
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, FO SRC
Color White
Footbed Material Cork
Material Leather
Sole Features Slip-resistant
View More Specs
Products 15
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15671468
View Documents Promotion Details
Abeba™
4090/34
34
81.95€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 25-04-2024
to see stock.
Add to basket
15681468
View Documents Promotion Details
Abeba™
4090/35
35
81.95€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 25-04-2024
to see stock.
Add to basket
15691468
View Documents Promotion Details
Abeba™
4090/36
36
81.95€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 25-04-2024
to see stock.
Add to basket
15601478
View Documents Promotion Details
Abeba™
4090/37
37
81.95€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 25-04-2024
to see stock.
Add to basket
15611478
View Documents Promotion Details
Abeba™
4090/38
38
81.95€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 25-04-2024
to see stock.
Add to basket
15621478
View Documents Promotion Details
Abeba™
4090/39
39
81.95€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 25-04-2024
to see stock.
Add to basket
15631478
View Documents Promotion Details
Abeba™
4090/40
40
81.95€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 25-04-2024
to see stock.
Add to basket
15641478
View Documents Promotion Details
Abeba™
4090/41
41
81.95€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 25-04-2024
to see stock.
Add to basket
15651478
View Documents Promotion Details
Abeba™
4090/42
42
81.95€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 25-04-2024
to see stock.
Add to basket
15661478
View Documents Promotion Details
Abeba™
4090/43
43
81.95€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 25-04-2024
to see stock.
Add to basket
15671478
View Documents Promotion Details
Abeba™
4090/44
44
81.95€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 25-04-2024
to see stock.
Add to basket
15681478
View Documents Promotion Details
Abeba™
4090/45
45
81.95€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 25-04-2024
to see stock.
Add to basket
15691478
View Documents Promotion Details
Abeba™
4090/46
46
81.95€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 25-04-2024
to see stock.
Add to basket
15601488
View Documents Promotion Details
Abeba™
4090/47
47
81.95€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 25-04-2024
to see stock.
Add to basket
15611488
View Documents Promotion Details
Abeba™
4090/48
48
81.95€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 25-04-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather
  • White
  • Slip resistance special rubber outsole sole
  • Leather coated cork footbed
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • 2 adjustable instep straps
  • velcro fastening
Specifications
Specifications
CE EN ISO 20347:2012 OB A, FO SRC
Cork
Slip-resistant
4090
White
Leather
Rubber