Learn More
Abeba™ Nature 4096 Shoes
Leather covered cork footbed is ESD dissipative.
Brand:  Abeba™ 4096/37
Additional Details : Weight : 0.78000kg
 View more versions of this product
Product Code. 15651488
  • 87.16€ / 1 pair
Estimated Shipment: 31-05-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Velours
  • Blue
  • Slip resistance special rubber outsole sole
  • Leather coated cork footbed
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • 2 adjustable instep straps
  • Buckle fastening
  • Metal-free
Specifications
Specifications
CE EN ISO 20347:2012 OB A, FO SRC
Cork
37
Rubber
Blue
Velour
Slip-resistant
4096