missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NCT 02
GREENER_CHOICE

Product Code. p-7223005
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18725496

Brand: Tocris Bioscience™ 7551/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Molecular glue; induces degradation of CCNK and CDK12

NCT 02 acts as a molecular glue that induces proteasomal degradation of CCNK and CDK12 in colorectal cancer spheroid cells in vitro (IC50 ≤ 50 μM). NCT 02 also induces apoptosis and the downregulation of genes involved in DNA damage repair.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6,7-Dihydro-N-(5-methyl-2-thiazolyl)-5 H-indeno[5,6-b]furan-3-acetamide
CAS 790245-61-3
Quantity 10 mg
Target Molecular Glues
Molecular Formula C17H16N2O2S
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.