missing translation for 'onlineSavingsMsg'
Learn More

Ndelta-Boc-L-ornithine, 98%

Product Code. 15429258
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15429258 5 g 5g
15419258 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15429258

Brand: Thermo Scientific Alfa Aesar H62774.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13650-49-2 | C10H20N2O4 | 232.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13650-49-2
Molecular Formula C10H20N2O4
Molecular Weight (g/mol) 232.28
MDL Number MFCD00037219
InChI Key GLZZMUULAVZVTA-ZETCQYMHSA-N
Synonym h-orn boc-oh, n-delta-boc-l-ornithine, boc-l-orn-oh, s-2-amino-5-tert-butoxycarbonyl amino pentanoic acid, ndelta-boc-l-ornithine, n-, a-boc-l-ornithine, l-ornithine, n5-1,1-dimethylethoxy carbonyl, n-epsilon-t-butyloxycarbonyl-l-ornithine, n∼5∼-tert-butoxycarbonyl-l-ornithine, s-2-amino-5-tert-butoxycarbonylamino-pentanoic acid
PubChem CID 6993436
IUPAC Name (2S)-2-amino-5-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
SMILES CC(C)(C)OC(=O)NCCCC(C(=O)O)N

Specifications

Quantity 5 g
Formula Weight 232.28
Percent Purity 98%
Chemical Name or Material Ndelta-Boc-L-ornithine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.