Learn More
Memmert™ Nearly Gastight Interior for UN30
Product Code. 17550096 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN30 Oven
Unit Size:
Each
Product Code. 17550096
Brand: Memmert™ UN30K2
Only null left
Add to basket
Add to basket
Interior nearly gastight after validation for combination with other options.
  • Technical clarification required for combination with other options
Specifications
For Use With (Equipment) UN30 Oven
Type Interior