Learn More
NEK11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA553516
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: DFGVSRLLMG SCDLATTLTG TPHYMSPEAL KHQGYDTKSD IWSLACILYE MCCMNHAFAG SNFLSIVLKI VEGDTPSLPE RYPKELNAIM ESMLNKNPSL RPSAIEILKI PYLDEQLQNL MC Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 82%.
NEK11 Polyclonal antibody specifically detects NEK11 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
NEK11 | |
Polyclonal | |
Unconjugated | |
Nek11 | |
4932416N14Rik; Nek11; never in mitosis A-related kinase 11; NIMA (never in mitosis gene a)- related kinase 11; NIMA (never in mitosis gene a)-related expressed kinase 11; NIMA related kinase 11; NIMA-related kinase 11; nimA-related protein kinase 11; Serine/threonine-protein kinase Nek11 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79858 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NG66 | |
Nek11 | |
Recombinant protein corresponding to Human NEK11 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only