missing translation for 'onlineSavingsMsg'
Learn More

Dr.Weigert™ Neodisher LaboClean FT

Product Code. 15701618
Change view
Click to view available options
Volume (Metric):
5 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
15701618 5 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15701618 Supplier Dr.Weigert™ Supplier No. 410033

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Neodisher LaboClean FT - Alkaline detergent, liquid concentrate. For automated cleaning of laboratory glassware in microbioly, virology, nuclear medecine, food industry and paint industry.

  • Alkaline detergent, liquid concentrate
  • With oxidizing effect
  • Free of surfactants
  • Neodisher Laboclean FT is specially suitable for the residue-free cleaning of glassware for cell and tissue cultures, pharmacy glass bottles and infusion flasks as well as glassware for microbiology and nuclear medicine.
  • Removes residues of culture media, residues of cell and tissue cultures, blood, proteins, radioactive contamination as well as other organic residues
  • Dosage between 2-4 mL/L

Specifications

Form Liquid
pH 11.8 to 12.2
Sterility Non-sterile
Composition <5 % amphoteric surfactants 5 - 15 % polycarboxylates
Concentration 2 to 5 mL/L
Container Type Canister
Content And Storage Store in a frost-free place at 0°C to 30°C. Usable for one year when stored as recommended.
Density 1.2 g/cm3 (20°C)
Description 220 x 240 x 140 mm
Viscosity <50 mPa.s (20°C)
Volume (Metric) 5 L
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.