Learn More
Euro Protection™ Neoprene Chemical Resistant Gloves
Product Code. p-8009266
Click to view available options
Size:
2X-Large, 11
Large, 9
Medium, 8
Small, 7
X-Large, 10
Unit Size:
10 pairs
Product Code. 12098255
Brand: Euro Protection™ 5307
Only null left
Add to basket
Add to basket
Wear Euro Protection™ Neoprene Gloves Neoprene so hands can be safe from a broad range of oils, acids, caustics and solvents. Neoprene synthetic rubber is less resistant to snags, punctures, abrasions and cuts than nitrile or natural rubber.
  • Black
  • Cotton flock lined
  • Grip pattern
Specifications
Color Black
Exterior Finish Grip Pattern
Material Polychloroprene, Rubber
Product Type Neoprene Gloves
Size Small, 7
Thickness (Metric) 0.7 mm
Lining Cotton flock lined
Quantity 10Pack
Length (Metric) 310 mm