missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Nephronophthisis 293T Cell Transient Overexpression Lysate

Product Code. 18319198 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.01mL
Product Code. Quantity unitSize
18319198 0.1 mL 0.01mL
1 options
This item is not returnable. View return policy

Product Code. 18319198

Brand: Novus Biologicals™ H00004867T02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Overexpression Lysate

TRUSTED_SUSTAINABILITY

Specifications

Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze-thaw cycles.
Research Category Cell Biology, Signal Transduction
Target Nephronophthisis
Synonym FLJ97602, JBTS4NPH1Juvenile nephronophthisis 1 protein, nephrocystin 1, nephrocystin-1, nephronophthisis 1 (juvenile), SLSN1
Gene Symbol NPHP1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.