missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Nepicastat hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Nepicastat hydrochloride is a potent and selective dopamine β-hydroxylase (DBH) inhibitor (IC50 = 9 nM) . Selective for DBH over 12 other enzymes. Decreases noradrenaline and increases dopamine levels in tissues and plasma in hypertensive rats. Prevents progressive left ventricular dysfunction in an in vivo heart failure model. Also potentiates the stimulus effects of cocaine without producing cocaine-like effects in rats. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 5-(Aminomethyl)-1-[(2S)-5,7-difluoro-1,2,3,4-tetrahydro-2-naphthalenyl]-1,3-dihydro-2H-imidazole-2-thione hydrochloride |
| CAS | 170151-24-3 |
| Quantity | 10 mg |
| Target | Hydroxylase Inhibitors |
| Molecular Formula | C14H15F2N3S.HCl |
| Formula Weight | 331.81 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction