missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Neratinib
GREENER_CHOICE

Product Code. 18768426
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. Quantity unitSize
18768426 50 mg 50mg
18768416 10 mg 10mg
2 options
This item is not returnable. View return policy

Product Code. 18768426

Brand: Tocris Bioscience™ 7371/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent HER2 receptor tyrosine kinase and EGFR kinase inhibitor

Neratinib is a potent, irreversible inhibitor of HER-2/ErbB2 receptor tyrosine kinase and EGFR kinase (IC50 = 59 nM and 92 nM, respectively). Neratinib reduces receptor autophosphorylation and proliferation of EGFR-dependent cancer cells, blocks cell-cycle progression and inhibits HER-2 signaling pathways. Neratinib suppresses proliferation of gefitinib -resistant non-small cell lung cancer cells expressing mutant EGFR. Also inhibits MST1 (IC50 = 37.7 nM). Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2 E)-N-[4-[[3-Chloro-4-(2-pyridinylmethoxy)phenyl]amino]-3-cyano-7-ethoxy-6-quinolinyl]-4-(dimethylamino)-2-butenamide
CAS 698387-09-6
Quantity 50 mg
Target EGF Receptor Inhibitors
Molecular Formula C30H29ClN6O3
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.