Learn More
Marienfeld Superior™ Counting Chamber with Double Net Ruling and V-Slash
Click to view available options
Quantity:
1Each
Unit Size:
Each
Description
- Precise measuring instrument for determination of the number of particles in liquids
- A V-slash facilitates the feeding of the capillary gap between chamber bottom and haemacytometer cover glass
- Made of special optical glass
- Equipped with two grids
- With 2 haemacytometer cover glasses 20x26 mm
- Individually packed in a transparent plastic box, 10 boxes in a carton
- For in-vitro diagnostic (IVD) applications according to IVD directive 98/79/EC, with CE-mark
Specifications
Specifications
| Product Type | Counting Chamber (Brightline) |
| Description | Neubauer-improved |
| Model | With V-slash |
| Quantity | 1Each |
| Depth (Metric) | 0.1 mm |
| Certifications/Compliance | CE Mark |
| Includes | Two cover glasses 20 x 26mm |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur