Learn More
Neuraminidase (NA) Recombinant Influenza A Virus Protein subtype H5N1 (A/Hubei/2011 (H5N1), N-His Tag
Neuraminidase (NA) Recombinant Influenza A Virus Protein subtype H5N1 (A/Hubei/2011 (H5N1), N-His Tag
Marque: Invitrogen 40018V07H25
Informations supplémentaires : Poids : 0.01000kg
Description
Neuraminidase (NA), recombinant Influenza A H5N1 (A/Hubei/2011) NA / Neuraminidase protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.This recombinant protein was expressed from a DNA sequence encoding the Influenza A H5N1 virus (A/Hubei/2011 (H5N1)) neuraminidase (His 36-Lys 448) fused to a polyhistidine tag at the N-terminus.
Activity: The activity of this protein has not been tested.
Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.
Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
This protein is manufactured by Sino Biological Inc.
Spécification
Protein assays and analysis | |
Lyophilized | |
47.9kDa | |
7.4 ±0.5 | |
≥90% by SDS-PAGE | |
–20°C | |
5 Pack(s) | |
Neuraminidase (NA) Recombinant Influenza A Virus Subtype H5N1 (A/Hubei/2011 (H5N1) Protein | |
NA | |
Other proteins | |
Neuraminidase (NA) Recombinant Influenza A Virus Subtype H5N1 (A/Hubei/2011 (H5N1) Protein | |
His Tag |
Lyophilized | |
Unknown | |
Neuraminidase (NA) Recombinant Influenza A Virus Subtype H5N1 (A/Hubei/2011 (H5N1) Protein | |
PAGE | |
5 x 5μg | |
For research use only. Not for use in diagnostic procedures | |
Viral proteins | |
5 Vials of protein in suspension buffer | |
Assay standard | |
Mammalian | |
Influenza |
For Research Use Only. Not for use in diagnostic procedures.