missing translation for 'onlineSavingsMsg'
Learn More

Neuraminidase (NA) Recombinant Influenza A Virus Protein subtype H5N1 (A/Hubei/2011 (H5N1), N-His Tag

Neuraminidase (NA) Recombinant Influenza A Virus Protein subtype H5N1 (A/Hubei/2011 (H5N1), N-His Tag

Marque:  Invitrogen 40018V07H250

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15286435

  • 1170.96€ / 250µg


nous consulter 056 260 260
Ajouter au panier

Description

Description

Neuraminidase (NA), recombinant Influenza A H5N1 (A/Hubei/2011) NA / Neuraminidase protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the Influenza A H5N1 virus (A/Hubei/2011 (H5N1)) neuraminidase (His 36-Lys 448) fused to a polyhistidine tag at the N-terminus.

Activity: The activity of this protein has not been tested.

Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

This protein is manufactured by Sino Biological Inc.

Spécification

Spécification

Protein assays and analysis
Lyophilized
47.9kDa
7.4 ±0.5
≥90% by SDS-PAGE
–20°C
5 Pack(s)
5 Vials of protein in suspension buffer
Assay standard
Mammalian
Influenza
Lyophilized
Unknown
Neuraminidase (NA) Recombinant Influenza A Virus Subtype H5N1 (A/Hubei/2011 (H5N1) Protein
PAGE
5 x 50μg
For research use only. Not for use in diagnostic procedures
Neuraminidase (NA) Recombinant Influenza A Virus Subtype H5N1 (A/Hubei/2011 (H5N1) Protein
NA
Other proteins
Neuraminidase (NA) Recombinant Influenza A Virus Subtype H5N1 (A/Hubei/2011 (H5N1) Protein
His Tag
Vidéos
Documents

For Research Use Only. Not for use in diagnostic procedures.