missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Neuropathiazol

Product Code. 18754099
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18754099

Brand: Tocris Bioscience™ 5186/10

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Selective inducer of neuronal differentiation in hippocampal neural progenitors

Neuropathiazol is a selective inducer of neuronal differentiation in cultured hippocampal neural progenitor cells (NPCs). Selectively suppresses astrocyte differentiation when induced by LIF, BMP2 and FBS compared to retinoic acid. Enhances maturation of NPC derived neurons.
TRUSTED_SUSTAINABILITY

Specifications

Purity >98%
Content And Storage -20°C
Quantity 10 mg
Product Type Neuropathiazol
Molecular Weight (g/mol) 338.42
Inhibitors Neuropathiazol

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.