Learn More
Thermo Scientific™ Neutralization Solution for MagJET Plasmid DNA Kit
Product Code. p-7184590 Shop All Thermo Scientific Products
Click to view available options
Quantity:
25 mL
90 mL
Unit Size:
25mL
90mL
Product Code. 13535230
Brand: Thermo Scientific™ R2601
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
The Thermo Scientific MagJET Plasmid DNA Kit is designed for fast and efficient purification of plasmid DNA from overnight cultures of E. coli cultures.
The Thermo Scientific MagJET Plasmid DNA Kit is designed for fast and efficient purification of plasmid DNA from overnight cultures of E. coli cultures. The kit utilizes magnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA is free of proteins, nucleases and other contaminants or inhibitors.
TRUSTED_SUSTAINABILITY
Specifications
Buffer Neutralization Solution
Product Line MagJET
Quantity 25 mL