Learn More
Neutrophil Elastase/ELA2 Antibody (950317R), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB91671RMFV610
523.20 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
Product Code. 18543049
  • 654.00€ / 0.10mL
Explore more special offers
Description
Description
Neutrophil Elastase/ELA2 Monoclonal antibody specifically detects Neutrophil Elastase/ELA2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF