missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ New Zealand Bovine Thrombin, High Specific Activity

Product Code. 17552896 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 KU
100 KU
5 KU
500 KU
Unit Size:
100ku
10ku
500ku
5ku
This item is not returnable. View return policy

Product Code. 17552896

Brand: MP Biomedicals™ 0218053910

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Manufactured in a state-of-the-art facility in New Zealand using a chromatographic extraction process, under Quality Systems that are ISO Certified and audited to cGMP Principles.

Thrombin is used throughout the diagnostics industry in a variety of coagulation assays, clotting factor tests and defibrination of blood or plasma for serum controls. It can also be applied to site specific cleavage of recombinant fusion proteins and in biochemical and medical research applications, specifically in hemostasis applications.

This bovine thrombin with high specific activity is manufactured in New Zealand from bovine plasma of New Zealand origin using prothrombin activated with thromboplastin extracted from bovine lung tissue. Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

  • Lyophilized in glass vials under vacuum to optimize product performance
  • Specific activity of >2000 NIH units/mg protein
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 KU
Regulatory Status Research Use Only
Species Bovine
Form Lyophilized
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.