missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ New Zealand Bovine Transferrin (Apo), Heat-treated

Product Code. p-7238942 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 17592716

Brand: MP Biomedicals™ 0215233480

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Bovine Transferrin is an iron-depleted product supplied as a heat-treated, lyophilized powder.

Bovine Transferrin (APO) is an iron-depleted product supplied as a heat-treated, lyophilized powder. Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

  • Frequently used as a nutrient in cell and microbial culture
  • Used as a media supplement in biopharmaceutical production
  • Useful transport factor in cell culture work to convey essential metabolites to the cells
  • Important factor when culturing cells in a defined medium
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Media Nutrient Supplement
Molecular Weight (g/mol) 76,000 to 81,000 g/mol
Quantity 100 mg
Regulatory Status Research Use Only
Species Bovine
Recombinant Native
Form Powder
Purity or Quality Grade Cell Culture Grade
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.