Learn More
LNI Swissgas NG CASTORE BASIC 180
Product Code. 17802029 Shop All LNI Swissgas Products
Click to view available options
Voltage:
100/115/220/230 V
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 17802029
Brand: LNI Swissgas 6920.52.180
Regulatory: This product is under eco-contribution tax.
Nitrogen generator with no dryer.
  • Needs source of dry and oil free air.
Specifications
Voltage 100/115/220/230 V
Type Nitrogen Gas Generator