Learn More
Thermo Scientific Chemicals Nickel Cover for Crucible 35925, 50ml
Product Code. 15401387
Click to view available options
Capacity (Metric):
5 mL
20 mL
25 mL
30 mL
35 mL
45 mL
50 mL
55 mL
75 mL
100 mL
250 mL
500 mL
1000 mL
Description:
For 35883 crucible
For 35889 crucible
For 35904 crucible
For 35906 crucible
For 35925 crucible
For 35927 crucible
For 35946 crucible
For 35969 crucible
For 35991 crucible
For 36019 crucible
For 36034 crucible
For 36040 crucible
For 36053 crucible
For 36057 crucible
For 36071 crucible
For 36084 crucible
Unit Size:
Each
Product Code. 15401387
Brand: Thermo Scientific Chemicals 035959.KT
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Description For 36053 crucible
Material Nickel
Capacity (Metric) 100 mL
Product Type Nickel Crucible Cover