Learn More
Thermo Scientific Chemicals Nickel Cover for Crucible 35925, 50ml
Click to view available options
Capacity (Metric):
5 mL
20 mL
25 mL
30 mL
35 mL
45 mL
50 mL
55 mL
75 mL
100 mL
250 mL
500 mL
1000 mL
Description:
For 35883 crucible
For 35889 crucible
For 35904 crucible
For 35906 crucible
For 35925 crucible
For 35927 crucible
For 35946 crucible
For 35969 crucible
For 35991 crucible
For 36019 crucible
For 36034 crucible
For 36040 crucible
For 36053 crucible
For 36057 crucible
For 36071 crucible
For 36084 crucible
Unit Size:
Each
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Description | For 35883 crucible |
| Material | Nickel |
| Capacity (Metric) | 20 mL |
| Product Type | Nickel Crucible Cover |
RUO – Research Use Only
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto