Learn More
Honeywell Fluka™ Nickel(II) Nitrate Hexahydrate, Honeywell™ Fluka™
Crystals or chunks
Brand: Honeywell Fluka™ 244074-500G
Additional Details : CAS : 13478-00-7 Poids : 0.50000kg Transport : UN number : 2725 Chem class : 5.1 Pack group : III
Packaging | Poly Bottle |
---|---|
Quantity | 500g |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
13478-00-7 | |
290.791 | |
AOPCKOPZYFFEDA-UHFFFAOYSA-N | |
61630 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Ni+2] |
H12N2NiO12 | |
MFCD00149805 | |
nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6 | |
nickel(2+);dinitrate;hexahydrate |
Specifications
Nickel(Ii) Nitrate Hexahydrate | |
H12N2NiO12 | |
MFCD00149805 | |
Moist Crystals. Chunks or Chips | |
AOPCKOPZYFFEDA-UHFFFAOYSA-N | |
nickel(2+);dinitrate;hexahydrate | |
61630 | |
Crystals Or Chunks | |
Poly Bottle | |
56°C |
13478-00-7 | |
Ni(NO3)2 · 6H2O | |
UN2725 | |
nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Ni+2] | |
290.791 | |
290.79g/mol | |
2.05g/mL at 25°C | |
Blue Green | |
500g |