missing translation for 'onlineSavingsMsg'
Learn More

Nickel Metal Sheet, Pure, about 0.5mm, Fisher Chemical™

Catalog Number p-7073332
Click to view available options
Quantity:
100 g
250 g
This item is not returnable. View return policy
7440-02-0
Ni
58.69
MFCD00011137 MFCD06798735
PXHVJJICTQNCMI-UHFFFAOYSA-N
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
935
CHEBI:28112
nickel
[Ni]
This item is not returnable. View return policy
7440-02-0
Ni
58.69
MFCD00011137 MFCD06798735
PXHVJJICTQNCMI-UHFFFAOYSA-N
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
935
CHEBI:28112
nickel
[Ni]
Melting Point 1455°C
Boiling Point 2732°C
Quantity 100 g
Formula Weight 58.7g/mol
Packaging Plastic container
Vapor Pressure 1mmHg 1810
Form Solid
compliance-icons compliance-icons
Product Identifier
  • Nickel, metal sheet
Signal Word
  • Danger
Hazard Category
  • Skin sensitiser Category 1
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H351-Suspected of causing cancer.
  • H372-Causes damage to organs.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'