Learn More
Bochem™ Nickel Plated Brass Pinch Cock
Product Code. 10425241
Click to view available options
Length (Metric):
30 mm
40 mm
50 mm
Unit Size:
Each
Pack of 10
Product Code. 10425241
Brand: Bochem™ 12540
Only null left
Add to basket
Add to basket
Used to control the flow of liquid through flexible tubing
  • With flap
  • Manufactured from Nickel Plated Brass
Specifications
Type Hoffman Pinchcock
Length (Metric) 50 mm