Learn More
Nimesulide, Thermo Scientific™
Highly selective cyclooxygenase-2 inhibitor which induces apoptosis
59.00€ - 231.99€
Chemical Identifiers
| CAS | 51803-78-2 |
|---|---|
| Molecular Formula | C13H12N2O5S |
| Molecular Weight (g/mol) | 308.308 |
| MDL Number | MFCD00079470 |
| InChI Key | HYWYRSMBCFDLJT-UHFFFAOYSA-N |
| Synonym | nimesulide, mesulid, n-4-nitro-2-phenoxyphenyl methanesulfonamide, flogovital, sulidene, nimed, 4-nitro-2-phenoxymethanesulfonanilide, nisulid, nimesulidum inn-latin, nimesulida inn-spanish |
| PubChem CID | 4495 |
| ChEBI | CHEBI:44445 |
| IUPAC Name | N-(4-nitro-2-phenoxyphenyl)methanesulfonamide |
| SMILES | CS(=O)(=O)NC1=C(C=C(C=C1)[N+](=O)[O-])OC2=CC=CC=C2 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15455879
|
Thermo Scientific Alfa Aesar
J63699.06 |
5g |
60.20€ 59.00€ / 5g Save 1.20€ 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15465879
|
Thermo Scientific Alfa Aesar
J63699.14 |
25g |
237.00€ 231.99€ / 25g Save 5.01€ 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble in water (<50 µg/ml), 1:10 DMSO:PBS (pH 7.2) (<200 µg/ml), ethanol (1 mg/ml), DMSO (15 mg/ml), DMF (15 mg/ml), chloroform, dichloromethane, acetone (freely soluble), and 1N NaOH.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
| 51803-78-2 | |
| 308.308 | |
| HYWYRSMBCFDLJT-UHFFFAOYSA-N | |
| 4495 | |
| N-(4-nitro-2-phenoxyphenyl)methanesulfonamide |
| C13H12N2O5S | |
| MFCD00079470 | |
| nimesulide, mesulid, n-4-nitro-2-phenoxyphenyl methanesulfonamide, flogovital, sulidene, nimed, 4-nitro-2-phenoxymethanesulfonanilide, nisulid, nimesulidum inn-latin, nimesulida inn-spanish | |
| CHEBI:44445 | |
| CS(=O)(=O)NC1=C(C=C(C=C1)[N+](=O)[O-])OC2=CC=CC=C2 |
Specifications
| 51803-78-2 | |
| MFCD00079470 | |
| 14,6548 | |
| Soluble in water (<50 μg/ml),1:10 DMSO:PBS (pH 7.2) (<200 μg/ml),ethanol (1mg/ml),DMSO (15mg/ml),DMF (15mg/ml),chloroform,dichloromethane,acetone (freely soluble),and 1N NaOH. | |
| CS(=O)(=O)NC1=C(C=C(C=C1)[N+](=O)[O-])OC2=CC=CC=C2 | |
| 308.308 | |
| CHEBI:44445 | |
| Nimesulide |
| C13H12N2O5S | |
| UN2811 | |
| nimesulide, mesulid, n-4-nitro-2-phenoxyphenyl methanesulfonamide, flogovital, sulidene, nimed, 4-nitro-2-phenoxymethanesulfonanilide, nisulid, nimesulidum inn-latin, nimesulida inn-spanish | |
| HYWYRSMBCFDLJT-UHFFFAOYSA-N | |
| N-(4-nitro-2-phenoxyphenyl)methanesulfonamide | |
| 4495 | |
| 308.31 |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P310-P330-P501c
H301
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 257-431-4
missing translation for 'rtecsNumber' : PB0970000
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.