Learn More
Analytik Jena US NIR Excitation Laser Modules
Analytik Jena US NIR Excitation Laser Modules are for use with UVP ChemStudio series
Brand:  Analytik Jena US 849-98-0123-01
Additional Details : Weight : 1.00000kg
Product Code. 15729769
  • 14560.00€ / Each
Estimated Shipment: 07-08-2024
to see stock.
Add to basket
Includes: Two emission filters
Description
Description
  • Excitation wavelengths of 660nm and 787nm.
  • Modules include 2 emission filters with wavelengths: 707-752nm and 810LP
Specifications
Specifications
1.5 in.
5 in.
5.08cm
2 in.
For use with UVP ChemStudio systems
12.7cm