Learn More
Nisin, 900 I.U./mg, Thermo Scientific™
Nisin, CAS # 1414-45-5, is a polycyclic peptide derived from and produced by Lactococcus lactis that shows antibacterial activity against gram-positive bacteria.
72.00€ - 590.00€
Chemical Identifiers
| CAS | 1414-45-5 |
|---|---|
| Molecular Formula | C143H230N42O37S7 |
| Molecular Weight (g/mol) | 3354.09 |
| MDL Number | MFCD00131724 |
| InChI Key | NVNLLIYOARQCIX-OAXMGIFASA-N |
| PubChem CID | 131847819 |
| SMILES | CCC(C)C1C(=O)NC(=C)C(=O)NC(C(=O)NC(CSCC(C(=O)N1)NC(=O)C(=CC)NC(=O)C(C(C)CC)N)C(=O)NC2C(SCC(NC(=O)CNC(=O)C3CCCN3C2=O)C(=O)NC(CCCCN)C(=O)NC4C(SCC(NC(=O)CNC(=O)C(NC(=O)C(NC(=O)C(NC(=O)CNC4=O)C)CC(C)C)CCSC)C(=O)NC(CC(=O)N)C(=O)NC(CCSC)C(=O)NC(CCCCN)C(=O)NC5C( |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15487699
|
Thermo Scientific Alfa Aesar
J66370.03 |
1g |
73.40€ 72.00€ / 1g Save 1.40€ 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15497699
|
Thermo Scientific Alfa Aesar
J66370.06 |
5g |
175.00€ 172.00€ / 5g Save 3.00€ 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15407709
|
Thermo Scientific Alfa Aesar
J66370.14 |
25g |
602.00€ 590.00€ / 25g Save 12.00€ 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identificadores de productos químicos
| 1414-45-5 | |
| 3354.09 | |
| NVNLLIYOARQCIX-OAXMGIFASA-N | |
| CCC(C)C1C(=O)NC(=C)C(=O)NC(C(=O)NC(CSCC(C(=O)N1)NC(=O)C(=CC)NC(=O)C(C(C)CC)N)C(=O)NC2C(SCC(NC(=O)CNC(=O)C3CCCN3C2=O)C(=O)NC(CCCCN)C(=O)NC4C(SCC(NC(=O)CNC(=O)C(NC(=O)C(NC(=O)C(NC(=O)CNC4=O)C)CC(C)C)CCSC)C(=O)NC(CC(=O)N)C(=O)NC(CCSC)C(=O)NC(CCCCN)C(=O)NC5C( |
| C143H230N42O37S7 | |
| MFCD00131724 | |
| 131847819 |
Especificaciones
| 1414-45-5 | |
| >110°C (230°F) | |
| MFCD00131724 | |
| 14,6564 | |
| NVNLLIYOARQCIX-OAXMGIFASA-N | |
| 3354.09 | |
| 3354.06 |
| Brown | |
| C143H230N42O37S7 | |
| 2967°C | |
| Soluble in water | |
| CCC(C)C1C(=O)NC(=C)C(=O)NC(C(=O)NC(CSCC(C(=O)N1)NC(=O)C(=CC)NC(=O)C(C(C)CC)N)C(=O)NC2C(SCC(NC(=O)CNC(=O)C3CCCN3C2=O)C(=O)NC(CCCCN)C(=O)NC4C(SCC(NC(=O)CNC(=O)C(NC(=O)C(NC(=O)C(NC(=O)CNC4=O)C)CC(C)C)CCSC)C(=O)NC(CC(=O)N)C(=O)NC(CCSC)C(=O)NC(CCCCN)C(=O)NC5C( | |
| 131847819 | |
| Nisin, 900 I.U./mg |
Seguridad y manipulación
missing translation for 'einecsNumber' : 215-807-5
missing translation for 'rtecsNumber' : QU3340000
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.