Learn More
Ansell™ NitraSafe™ 28-329 Series Nitrile Heavy Weight Gloves
Product Code. 12610812 Shop All Ansell Products
Click to view available options
Size:
7
8
9
10
Unit Size:
12 pairs
Product Code. 12610812
Brand: Ansell™ 28329_7
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Combines the outstanding protection of Kevlar™-reinforced construction with the grip and flexibility of Ansell’s advanced nitrile coating technology. The NitraSafe™ range stands for safe handling of oily heavy metal parts, while the NitraSafe™ Foam combines good grip and comfort.
  • Cut and sewn construction
  • Cotton Jersey Kevlar™ liner
  • Nitrile coating
  • Palm-coated finish
  • Nitragold™ cuff
TRUSTED_SUSTAINABILITY
Specifications
Back Material Nitrile
Certifications/Compliance CE Mark 0493, Standard Category III EN 388 : 4331
Cuff Material Nitrile
Coating Material Nitrile
Color Black
Cut Resistance Rating Level C
Hand Specific
Latex Free Yes
Material Nitrile
Product Type Heavyweight Gloves
Size 7
Glove Cuff Style Nitragold™
Liner Material Cotton Jersey Dupont™ Kevlar™ aramid fiber
Lining Lined
Palm Type Coated-palm
Quantity 72 Pairs
Resistant To Greater resistance to cuts and abrasion