Learn More
Ansell™ NitraSafe™ 28-329 Series Nitrile Heavy Weight Gloves
Shop All Ansell Products
Click to view available options
Size:
7
8
9
10
Unit Size:
12 pairs
Description
- Cut and sewn construction
- Cotton Jersey Kevlar™ liner
- Nitrile coating
- Palm-coated finish
- Nitragold™ cuff
Specifications
Specifications
| Back Material | Nitrile |
| Certifications/Compliance | CE Mark 0493, Standard Category III EN 388 : 4331 |
| Cuff Material | Nitrile |
| Coating Material | Nitrile |
| Color | Black |
| Cut Resistance Rating | Level C |
| Hand | Specific |
| Latex Free | Yes |
| Material | Nitrile |
| Product Type | Heavyweight Gloves |
| Show More |