missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitril™ 3000 X-Long, Non-Sterile, Nitrile Examination Gloves
Description
- AQL 1,5
- PPE Category III
- Class I Medical
- Extra long length
- Ambidextrous
- Non-sterile
- Powder free and latex free
- Textured
- Beaded cuff
Specifications
Specifications
| Certifications/Compliance | AQL 1,5, PPE Cat III, Class I Medical, Food compliant, EN 455-1/-2/-3/-4, EN 374-1/-2/-4/-5, EN 16623-1, EN 420, EN ISO 13495:2016, 93/42/EEC |
| Color | Blue |
| Disposable | Single-use |
| Material | Nitrile |
| Size | Medium |
| Model | 1282M |
| Length (Metric) | 29 cm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?