Learn More
Lovibond™ Nitrite/Nitrite-N Tube Test for SpectroDirect Spectrophotometer
Reactor Necessary - Tubes must be heated to 100°C
Brand:  Lovibond™ 2419018
Additional Details : Weight : 0.50000kg
Product Code. 10404922
  • 87.40€ / Each
Estimated Shipment: 30-04-2024
to see stock.
Specifications
24 Tests
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • NITRITE/NITRITE-N TUBE TEST 540 - 545 NM 40
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P313-Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : adipic acid, sulphanilic acid