missing translation for 'onlineSavingsMsg'
Learn More

Nitroterephthalic acid, 99%, Thermo Scientific Chemicals

Catalog Number p-7027959
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
610-29-7
C8H5NO6
211.129
MFCD00007141
QUMITRDILMWWBC-UHFFFAOYSA-N
nitroterephthalic acid, 2-nitroterephthalic acid, 2-nitrobenzene-1,4-dicarboxylic acid, 1,4-benzenedicarboxylic acid, 2-nitro, terephthalic acid, nitro, 2-nitroterephthalicacid, 2-nitroterephthalate, 5-nitroterephthalic acid, acmc-1au2u
69122
C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])C(=O)O
This item is not returnable. View return policy
610-29-7
C8H5NO6
211.129
MFCD00007141
QUMITRDILMWWBC-UHFFFAOYSA-N
nitroterephthalic acid, 2-nitroterephthalic acid, 2-nitrobenzene-1,4-dicarboxylic acid, 1,4-benzenedicarboxylic acid, 2-nitro, terephthalic acid, nitro, 2-nitroterephthalicacid, 2-nitroterephthalate, 5-nitroterephthalic acid, acmc-1au2u
69122
C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 270°C to 272°C
Quantity 5 g
Formula Weight 211.13
Percent Purity 99%
Chemical Name or Material Nitroterephthalic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'