missing translation for 'onlineSavingsMsg'
Learn More

Nocodazole, 98%, Thermo Scientific Chemicals

Catalog Number p-175686
Click to view available options
Quantity:
10 mg
50 mg
Packaging:
Glass bottle
31430-18-9
C14H11N3O3S
301.32
MFCD00005588
KYRVNWMVYQXFEU-UHFFFAOYSA-N
nocodazole, oncodazole, nocodazol, nocodazolum, nocidazole, nocodazole usan:inn, nocodazol inn-spanish, nocodazolum inn-latin, unii-sh1wy3r615, methyl 5-2-thenoyl-2-benzimidazolecarbamate
4122
CHEBI:34892
methyl N-[6-(thiophene-2-carbonyl)-1H-benzimidazol-2-yl]carbamate
COC(=O)NC1=NC2=C(N1)C=C(C=C2)C(=O)C3=CC=CS3
31430-18-9
C14H11N3O3S
301.32
MFCD00005588
KYRVNWMVYQXFEU-UHFFFAOYSA-N
nocodazole, oncodazole, nocodazol, nocodazolum, nocidazole, nocodazole usan:inn, nocodazol inn-spanish, nocodazolum inn-latin, unii-sh1wy3r615, methyl 5-2-thenoyl-2-benzimidazolecarbamate
4122
CHEBI:34892
methyl N-[6-(thiophene-2-carbonyl)-1H-benzimidazol-2-yl]carbamate
COC(=O)NC1=NC2=C(N1)C=C(C=C2)C(=O)C3=CC=CS3

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 300.0°C to 305.0°C
Color Brown-Orange to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 mg
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dmso,dmf,ethanol,chloroform,acetone
Formula Weight 301.32
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Nocodazole, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Nocodazole
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H361d-Suspected of damaging the unborn child.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'