missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Non-human Primate IL-4 ELISA Kit

Product Code. 18356352 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit (1 Plate)
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18356352

Brand: Novus Biologicals™ NBP311724

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 3.16 - 1000 pg/mL
Assay Sensitivity 3 pg/mL
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 3565
Gene Symbol IL4
Quantity 1 Kit (1 Plate)
Research Discipline Adaptive Immunity, Asthma, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym B cell growth factor 1, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL-4B_cell stimulatory factor 1, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra
Target Non-human Primate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.