missing translation for 'onlineSavingsMsg'
Learn More

Nonyl aldehyde, 95%, Thermo Scientific Chemicals

Catalog Number p-175739
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
124-19-6
C9H18O
142.24
MFCD00007030
GYHFUZHODSMOHU-UHFFFAOYSA-N
pelargonaldehyde, 1-nonanal, nonanaldehyde, nonyl aldehyde, n-nonaldehyde, nonylic aldehyde, nonylaldehyde, nonaldehyde, n-nonanal, 1-nonaldehyde
31289
CHEBI:84268
nonanal
CCCCCCCCC=O
This item is not returnable. View return policy
124-19-6
C9H18O
142.24
MFCD00007030
GYHFUZHODSMOHU-UHFFFAOYSA-N
pelargonaldehyde, 1-nonanal, nonanaldehyde, nonyl aldehyde, n-nonaldehyde, nonylic aldehyde, nonylaldehyde, nonaldehyde, n-nonanal, 1-nonaldehyde
31289
CHEBI:84268
nonanal
CCCCCCCCC=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8270g/mL
Boiling Point 93.0°C (23.0 mmHg)
Flash Point 63°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)7CHO
Refractive Index 1.4230 to 1.4270
Quantity 100 mL
Beilstein 01,708
Specific Gravity 0.827
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol
Formula Weight 142.24
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Nonyl aldehyde, 95%
Show More Show Less
compliance-icons
Product Identifier
  • Nonyl aldehyde
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'