Learn More
(+)-Nootkatone, crystalline, 98+%
Description
(+)-Nootkatone, crystalline is used as an effective repellent in mosquitos, bed bugs, lone star ticks and head lice. It is also used as a food additive and flavoring material in beverages. It finds application in fragrances, cosmetics and pharmaceuticals. Further, it is used as a flavor material in beverage, frozen dairy and baked good applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
(+)-Nootkatone, crystalline is used as an effective repellent in mosquitos, bed bugs, lone star ticks and head lice. It is also used as a food additive and flavoring material in beverages. It finds application in fragrances, cosmetics and pharmaceuticals. Further, it is used as a flavor material in beverage, frozen dairy and baked good applications.
Solubility
Slightly soluble in ethanol and chloroform. Partly soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 34°C to 36°C |
| Density | 0.997 |
| Boiling Point | 125°C (0.5 mmHg) |
| Flash Point | 99°C (210°F) |
| Quantity | 25 g |
| Beilstein | 4676969 |
| Solubility Information | Slightly soluble in ethanol and chloroform. Partly soluble in water. |
| Optical Rotation | +185° (c=1 in Chloroform) |
| Formula Weight | 218.34 |
| Percent Purity | ≥98% |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.