missing translation for 'onlineSavingsMsg'
Learn More

Norethindrone, TRC

Product Code. 30422560
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Product Code. Quantity
30422560 100 mg
30409828 1 g
30421959 250 mg
3 options
This item is not returnable. View return policy

Product Code. 30422560

Brand: Toronto Research Chemicals TRCN676000100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-22-4
Formula Weight 298.1933
InChI Formula InChI=1S/C20H26O2/c1-3-20(22)11-9-18-17-6-4-13-12-14(21)5-7-15(13)16(17)8-10-19(18,20)2/h1,12,15-18,22H,4-11H2,2H3/t15-,16+,17+,18-,19-,20-/m0/s1
IUPAC Name (8R,9S,10R,13S,14S,17R)-17-ethynyl-17-hydroxy-13-methyl-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one
Molecular Formula C20 H26 O2
Molecular Weight (g/mol) 298.42
Percent Purity >95%
SMILES C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1CC[C@@]2(O)C#C
Synonym Norethisterone Acetate Imp. A (EP), Levonorgestrel Imp. U (EP), (17α)-17-Hydroxy-19-Norpregn-4-en-20-yn-3-one, 17-Hydroxy-19-nor-17α-pregn-4-en-20 yn-3-one, 17α-Ethinyl-17β-hydroxy-δ4-estren-3-one, 17α-Ethinyl-19-nortestosterone, 17α-Ethinylestr-4-en-17β-ol-3-one, 17α-Ethynyl-17-hydroxy-4-estrene-3-one, 17α-Ethynyl-17-hydroxyest-4-en-3-one, 17α-Ethynyl-17-hydroxyestr-4-en-3-one, 17α-Ethynyl-17β-hydroxy-19-norandrost-4-en-3-one, 17α-Ethynyl-17β-hydroxyestr-4-en-3-one, 17α-Ethynyl-19-nor-androst-4-en-17β-ol-3-one, 17α-Ethynyl-19-nortestosterone, 17α-Ethynyl-3-oxo-4-estren-17β-ol, 17β-Hydroxy-17α-ethynylestr-4-en-3-one, 19-Nor-17α-ethynyl-17β-hydroxy-4-androsten-3-one, 19-Nor-17α-ethynylandrosten-17β-ol-3-one, 19-Nor-17α-ethynyltestosterone, 19-Norandrost-4-en-3-one, 17α-ethynyl-17β-hydroxy-, 19-Nortestosterone, 17-ethynyl-, Anovule, Conludaf, Conludag, Estr-4-ene-17α-ethynyl-17β-ol-3-one, Ethinylnortestosterone, Ethynylnortestosterone, Gestest, Menzol, Micronett, Micronor, Micronovum, Mini-Pe, Mini-pill, NSC 9564, Nor-QD, Noralutin, Norcolut, Norethindrone, Norethisteron, Norethisterone, Norethynodrone, Noretisterone Dak, Norfor, Norgestin, Norluten, Norlutin, Norluton, Normapause, Norpregneninolone, Primolut N, Proluteasi, SC 4640, Triella, Utovlan
Chemical Name or Material Norethisterone
Grade HPLC
Show More Show Less

Specifications

Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.