Learn More
Masterflex™ L/S™ Norprene™ High-Pressure Pump Tubing
Click to view available options
Tubing Size:
15
16
Unit Size:
Each
Description
- Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
Specifications
Specifications
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 3.1 mm |
| Material | Norprene |
| Overall Length (Metric) | 7.6 m |
| Min. Operating Temperature (Metric) | -50°C |
| Tubing Size | 16 |
| Hose Barb | 1/8in. |
| Height (Metric) | 8.8 m |
| Maximum Pressure | 125 psi |
| Vacuum Level | 660 mm |
| Show More |