Learn More
Masterflex™ L/S™ Norprene™ High-Pressure Pump Tubing
Product Code. p-9016007
Click to view available options
Tubing Size:
15
16
Unit Size:
Each
Product Code. 11884242
Brand: Masterflex™ 0650415
Only null left
Add to basket
Add to basket
Designed to pump at higher pressures
  • Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
Specifications
Autoclavable Autoclavable
Diameter (Metric) Inner 4.8 mm
Material Norprene
Overall Length (Metric) 7.5 m
For Use With L/S high-performance pump head models 77250-62 and 77250-82 only
For Use With (Application) high pressure applications
Tubing Size 15