missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ North™ AK Cleanroom Rubber Gloves

Natural rubber latex cleanroom chemical-resistant gloves

Brand:  Honeywell™ AK1815O/9

Additional Details : Weight : 25.00000kg

 View more versions of this product

Product Code. 14871511

  • 377.88€ / Pack of 100


Estimated Shipment
10-06-2020
Add to basket

Alert:

These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.


Description

Description

  • 100% natural rubber latex has excellent dexterity and elasticity
  • Powder-free manufacturing process reduces particulate and extractable contamination
  • Diamond embossed palm provides added grip on wet surfaces
  • Cleanroom packaged to protect gloves from airborne contamination
  • Double bagged with cleanroom compatible packaging and labeling materials
  • Size embossed on gloves so there is no ink to contaminate process
  • Suitable for up to class 100 cleanroom

Order Info

Each pair individually packaged in a polybag.

Compliance

Testing: IEST-RP-CC005.3 (modified) particulate and extractables; ASTM™ 739 permeation.

Specifications

Specifications

Safety
Rubber
Powder-free
38 cm
Beaded/Rolled
9
Orange
Videos
Documents