missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NovaBright™ Phospha-Light™ EXP Assay Kit for SEAP (Secreted Placental Alkaline Phosphatase) Reporter Gene Detection

Chemiluminescent reporter gene assay for the ultrasensitive detection of secreted placental alkaline phosphatase or the non-secreted placental isozyme PLAP.

424.00€ - 1614.00€

Specifications

Product Type Phosphatase Assay
Detection Method Enhanced Chemiluminescence
For Use With (Equipment) Luminometer (Microplate)
Content And Storage Component solution containing CSPD™ substrate and chemiluminescence enhancer (Store at 2°C to 6°C), placental alkaline phosphatase positive control (Store at ≤ -20°C)
High-throughput Compatibility High-throughput Compatible
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10600550
View Documents Promotion Details
Invitrogen™
N10577
192 assays
424.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 10-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
10487308
View Documents Promotion Details
Invitrogen™
N10578
960 assays
1614.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 10-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sensitive detection of alkaline phosphatase reporter enzyme enables a large number of applications in many areas of life science research, including gene expression, viral function assays, vaccine development, development of viral vectors and gene delivery methods for gene therapy, in vivo gene expression monitoring and novel cellular functional assays. This SEAP assay is one of the easiest and fastest methods for optimizing transfection efficiency. This assay has been used for a wide variety of viral functional assays, including viral gene expression assays, viral replication, viral fusogenicity, virus neutralization and viral-mediated cell-cell fusion, and viral infectivity. Use of the SEAP reporter protein is very enabling for in vivo reporter gene assays, by assaying serum samples from transgenic, transfected or viral vector-infected animals. The NovaBright reporter gene assay system has been used to measure SEAP levels in sera from transgenic or transfected whole animals, including mouse, rat, marmoset, monkey and pig sera, and in chicken egg allantoic fluid. The mouse SEAP protein (mSEAP) has recently been developed for improved SEAP protein stability in transgenic mice, and the NovaBright SEAP assay has been used for sensitive detection of mSEAP . In addition to reporter gene (gene expression) applications, the assay system is used to measure SEAP as a functional reporter for receptor-ligand binding assays with a SEAP-ligand chimera, protease-mediated secretion, and for secretion pathway activity. The NovaBright assay system can be used for the cellular measurement of non-placental alkaline phosphatase as a biomarker.

This kit provides a much simpler procedure than other assays. The assay is homogenous and requires only two solutions both of which are supplied ready-to-use. Prolonged glow light emission kinetics provides a stable light signal for read-time flexibility. The assay signal is measured in a simple luminometer without injectors. SEAP is a reporter protein that is secreted into the cell culture medium and detected by testing aliquots of the medium. Since SEAP is released into the medium, cell lysis is unnecessary. However, the assay also allows the detection placental alkaline phosphase (PLAP) isoenzyme following cell lysis.

  • Second generation chemiluminescent reporter gene assay
  • Simple, fast protocol, high throughput-compatible
  • Ultrasensitive detection of secreted placental alkaline phosphatase
  • Compatibility with automated platforms with trigger solution/positions already in place
Specifications
  • Detection Method: Chemiluminescent
  • For Use With Equipment: Luminometer (Microplate)
  • Format: 96-well plate
  • High Throughput Compatibility: High Throughput-Compatible
  • Readout: End Point
  • Substrate: CSPD
  • Substrate Properties: Chemical Substrate
  • Substrate Type: AP (Alkaline Phosphatase) Substrate
  • Target Enzyme: Placental Alkaline Phosphatase
Specifications

Specifications

Phosphatase Assay
Luminometer (Microplate)
High-throughput Compatible
AP (Alkaline Phosphatase) Substrate
Placental Alkaline Phosphatase
NovaBright™
Enhanced Chemiluminescence
Component solution containing CSPD™ substrate and chemiluminescence enhancer (Store at 2°C to 6°C), placental alkaline phosphatase positive control (Store at ≤ -20°C)
CSPD™
Chemical Substrate
96-well Plate
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.