Learn More
Invitrogen™ Novex™ IL23 Heterodimer Human Antibody Pair
Concentrated
Brand: Invitrogen™ CHC2493
Includes: Antibody Pair Kit contains matched, pre-titred and fully optimized Capture (coating) and Detection Antibodies, 3 vials of Recombinant Standard, Streptavidin-HRP concentrate and technical data sheet.
Requires : Buffer reagents required to complete reaction available separately (Cat. No. CNB0011).
Description
Antibody Pair Kit comes with matched antibody pairs to detect and quantify protein levels of human IL-23 heterodimer.
Also available is Buffer Kit for Antibody Pairs that saves time with premade, easy-to-use buffers and solutions optimized for use with Invitrogen Antibody Pairs. The Assay Buffer can be used as a blocking reagent for ELISA plates as well as a diluent for ELISA standards and samples, detection antibody, and HRP conjugate.
Build Your Own Immunoassay, Cell Analysis, Immunoassays, Ready-To-Use Immunoassay
Order Info
Shipping Condition: Wet Ice
Specifications
| NM_016584.2, NP_057668.1 | |
| IL-23, IL-23A, IL23P19, MGC79388, P19, SGRF | |
| IL23A |